Learn More
Multiroir™ Crystal Polystyrene Minimax Storage Boxes
Click to view available options
Dimensions (L x W x H):
178 x 45 mm (L x H)
228 x 178 x 68 mm
228 x 178 x 72 mm
315 x 230 x 70 mm
315 x 45 mm (L x H)
Unit Size:
Each
Description
- Transparent as glass
- Delivered complete with separator kit
- Various capacities
Specifications
Specifications
| Color | Transparent |
| Material | Polystyrene |
| Shape | Rectangular |
| Product Type | Storage Box |
| Quantity | 6 Each |
| Dimensions (L x W x H) | 228 x 178 x 72 mm |
| Length (Metric) | 228 mm |
| Height (Metric) | 72 mm |