missing translation for 'onlineSavingsMsg'
Learn More

CRYZL1 Antibody (OTI2F9), DyLight 350, Novus Biologicals™

Product Code. 18190214 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18190214 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18190214 Supplier Novus Biologicals Supplier No. NBP271522UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRYZL1 Monoclonal specifically detects CRYZL1 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYZL1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2F9
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias crystallin, zeta (quinone reductase)-like 1, human quinone oxidoreductase homolog-1104P11QOH-1quinone oxidoreductase-like protein 1, Protein 4P11, Quinone oxidoreductase homolog 1, quinone reductase-like 1, Zeta-crystallin homolog
Gene Symbols CRYZL1
Host Species Mouse
Immunogen Full length recombinant protein of human CRYZL1 (NP_665857) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9946.0
Target Species Human, Mouse, Rat, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.