missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYZL1 Antibody (OTI2F9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02012
This item is not returnable.
View return policy
Description
CRYZL1 Monoclonal specifically detects CRYZL1 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CRYZL1 | |
| Monoclonal | |
| 1.08 mg/mL | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| CRYZL1 | |
| Full length human recombinant protein of human CRYZL1(NP_665857) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 9946 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI2F9 | |
| Unconjugated | |
| crystallin, zeta (quinone reductase)-like 1, human quinone oxidoreductase homolog-1104P11QOH-1quinone oxidoreductase-like protein 1, Protein 4P11, Quinone oxidoreductase homolog 1, quinone reductase-like 1, Zeta-crystallin homolog | |
| Mouse | |
| 38.5 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Primate | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction