Learn More
CRYZL1 Mouse anti-Human, Clone: OTI2F9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504659
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.Specifications
| CRYZL1 | |
| Monoclonal | |
| Unconjugated | |
| CRYZL1 | |
| 4P11, QOH-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9946 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2F9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O95825 | |
| CRYZL1 | |
| Full length human recombit protein of human CRYZL1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.