missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CSB Antibody [Allophycocyanin], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-61070APC
This item is not returnable.
View return policy
Description
CSB Polyclonal antibody specifically detects CSB in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Specifications
| CSB | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative) | |
| APC | |
| ARMD5, ATP-dependent helicase ERCC6, CKN2, Cockayne syndrome group B protein, Cockayne syndrome protein CSB, COFS, COFS1, CSBCockayne syndrome B protein, DNA excision repair protein ERCC-6, EC 3.6.1, EC 3.6.4.-, excision repair cross-complementing rodent repair deficiency, complementationgroup 6, RAD26, Rad26 homolog | |
| The immunogen recognized by this antibody maps to a region between residue 1161 and 1211 of human Cockayne syndrome protein B (excision repair cross-complementing rodent repair deficiency, complementation group 6) using the numbering given in entry NP_000115.1 (GeneID 2074). | |
| 0.1 mL | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2074 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction