missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSB Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61070B
This item is not returnable.
View return policy
Description
CSB Polyclonal specifically detects CSB in Human samples. It is validated for Western Blot.
Specifications
| CSB | |
| Polyclonal | |
| Western Blot | |
| ARMD5, ATP-dependent helicase ERCC6, CKN2, Cockayne syndrome group B protein, Cockayne syndrome protein CSB, COFS, COFS1, CSBCockayne syndrome B protein, DNA excision repair protein ERCC-6, EC 3.6.1, EC 3.6.4.-, excision repair cross-complementing rodent repair deficiency, complementationgroup 6, RAD26, Rad26 homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ERCC6 | |
| The immunogen recognized by this antibody maps to a region between residue 1161 and 1211 of human Cockayne syndrome protein B (excision repair cross-complementing rodent repair deficiency, complementation group 6) using the numbering given in entry NP_000115.1 (GeneID 2074). | |
| 0.1 mL | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2074 | |
| Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only