missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CSDE1 Antibody (SR4301), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 316.00 - € 382.00
Specifications
| Antigen | CSDE1 |
|---|---|
| Clone | SR4301 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041667
|
Novus Biologicals™
NBP3-27839-50ul |
50 μL |
€ 316.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041672
|
Novus Biologicals™
NBP3-27839-100ul |
100 μL |
€ 382.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CSDE1 Monoclonal antibody specifically detects CSDE1 in Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| CSDE1 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 7812 | |
| IgG | |
| Affinity purified |
| SR4301 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| cold shock domain containing E1, RNA-binding, cold shock domain-containing protein E1, D1S155EUNRRP5-1000E10.3, DKFZp779B0247, DKFZp779J1455, FLJ26882, KIAA0885, N-ras upstream gene protein, NRAS-related, NRU, Protein UNR, upstream of NRAS | |
| A synthesized peptide derived from human CSDE1 (Uniprot #: O75534) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title