missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSE1L/CAS/Exportin-2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79803F
This item is not returnable.
View return policy
Description
CSE1L/CAS/Exportin-2 Polyclonal antibody specifically detects CSE1L/CAS/Exportin-2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CSE1L/CAS/Exportin-2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CASMGC117283, Cellular apoptosis susceptibility protein, chromosome segregation 1 (yeast homolog)-like, Chromosome segregation 1-like protein, CSE1, CSE1 chromosome segregation 1-like (yeast), CSE1L, Exp2, exportin-2, Importin-alpha re-exporter, MGC130037, XPO2MGC130036 | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Chromosome Segregation-1 using the numbering given in entry NP_001307.2 (GeneID 1434). | |
| 0.1 mL | |
| Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers | |
| 1434 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction