missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSE1L Monoclonal Antibody (OTI2C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812194S
This item is not returnable.
View return policy
Description
CSE1L Monoclonal antibody specifically detects CSE1L in Human samples. It is validated for Western BlotSpecifications
| CSE1L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y6R0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9253 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI2C5 | |
| Unconjugated | |
| Numbl | |
| 2610100P18Rik; AA407533; Capts; CAS; Cellular apoptosis susceptibility protein; chromosome segregation 1 like; chromosome segregation 1-like (S. cerevisiae); chromosome segregation 1-like protein; CSE1; CSE1 chromosome segregation 1 like; CSE1 chromosome segregation 1-like; CSE1 chromosome segregation 1-like (yeast); CSE1L; Exp2; exportin-2; Importin-alpha re-exporter; MGC117283; MGC130036; MGC130037; XPO2 | |
| Human recombinant protein fragment corresponding to amino acids 1-225 of human NUMBL produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction