missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSF1 Mouse anti-Human, Clone: OTI2D6, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806568S
This item is not returnable.
View return policy
Description
CSF1 Monoclonal antibody specifically detects CSF1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CSF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P09603 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1435 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D6 | |
| Unconjugated | |
| Csf1 | |
| C87615; colony stimulating factor 1; colony stimulating factor 1 (macrophage); colony-stimulating factor-1 splice variant; CSF1; CSF-1; Csfm; H-MCSF; lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; macrophage colony-stimulating factor; macrophage colony-stimulating factor 1; macrophage colony-stimulating factor beta; macrophage-colony stimulating factor alpha; MCSF; MCSF alpha; MCSFBETA; MGC31930; M-MCSF; op; osteopetrosis; Processed macrophage colony-stimulating factor 1 | |
| Human recombinant protein fragment corresponding to amino acids 223-496 of human CSF1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction