missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSF2RA Monoclonal Antibody (OTI3E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812523
This item is not returnable.
View return policy
Description
CSF2RA is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2, a cytokine which controls the production, differentiation, and function of granulocytes and macrophages. The encoded protein is a member of the cytokine family of receptors. This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes. Multiple transcript variants encoding different isoforms have been found for this gene, with some of the isoforms being membrane-bound and others being soluble.Specifications
| CSF2RA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| CD116, CDw116, CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF-R-alpha, GMCSFR, GMR, SMDP4, alphaGMR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1438 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3E6 | |
| Unconjugated | |
| CSF2RA | |
| CSF2RA | |
| Human recombit protein fragment corresponding to amino acids 23-320 of human CSF2RA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction