missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSGALNACT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599746
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CSGALNACT1.
CSGALNACT1 transfers 1,4-N-acetylgalactosamine (GalNAc) from UDP-GalNAc to the non-reducing end of glucuronic acid (GlcUA). CSGALNACT1 required for addition of the first GalNAc to the core tetrasaccharide linker and for elongation of chondroitin chains. CSGALNACT1 is important role in chondroitin chain biosynthesis in cartilage.
Specifications
| CSGALNACT1 | |
| Polyclonal | |
| Unconjugated | |
| CSGALNACT1 | |
| 4732435N03Rik; beta1,4 N-acetylgalactosaminyltransferase; beta4GalNAcT; Beta4GalNAcT-1; Chgn; chondroitin beta1,4 N-acetylgalactosaminyltransferase; Chondroitin beta-1,4-N-acetylgalactosaminyltransferase 1; chondroitin sulfate N-acetylgalactosaminyltransferase 1; CSGALNACT1; CsGalNAcT-1; Galnact1; glucuronylgalactosylproteoglycan 4-beta-N- acetylgalactosaminyltransferase; RGD1307618; UNQ656/PRO1287 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55790 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TDX6 | |
| CSGALNACT1 | |
| A synthesized peptide derived from human CSGALNACT1(Accession Q8TDX6), corresponding to amino acid residues G311-I361. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction