missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ csgD Polyclonal Antibody

Product Code. 17959802
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17959802 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17959802 Supplier Invitrogen™ Supplier No. PA5144509

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

csgD Polyclonal Antibody for Western Blot, ELISA

Necessary for transcription of the csgAB operon. May have the capability to respond to starvation and/or high cell density by activating csgBA transcription (By similarity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen csgD
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.559 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene csgD
Gene Accession No. O54294
Gene Alias Probable csgAB operon
Gene Symbols csgD
Host Species Rabbit
Immunogen Recombinant Salmonella typhimurium (strain LT2/SGSC1412/ATCC 700720) csgD protein (1-216aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1252660
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.