missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSH1 Mouse anti-Human, Clone: OTI2A2, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506813S
This item is not returnable.
View return policy
Description
CSH1 Monoclonal antibody specifically detects CSH1 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| CSH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P0DML2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1442 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| OTI2A2 | |
| Unconjugated | |
| CSH1 | |
| Choriomammotropin; chorionic somatomammotropin A; chorionic somatomammotropin hormone 1; chorionic somatomammotropin hormone 1 (placental lactogen); Chorionic somatomammotropin hormone 2; chorionic somatomammotropin-1; CS-1; CSA; CSH1; CSH2; CSMT; GHB3; growth hormone B3; hCS-1; hCS-A; hPL; Lactogen; PL; placental lactogen | |
| Full length human recombinant protein of human CSH1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction