missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CSK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29909
This item is not returnable.
View return policy
Description
CSK Polyclonal antibody specifically detects CSK in Human samples. It is validated for Immunoprecipitation
Specifications
| CSK | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
| Between 375 and 425 | |
| 100 μg | |
| Cell Cycle and Replication, Signal Transduction | |
| 1445 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction