missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSK Mouse anti-Human, Clone: 7C6, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59500-50UL
This item is not returnable.
View return policy
Description
CSK Monoclonal specifically detects CSK in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CSK | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 7C6 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| CSK | |
| Recombinant human CSK (1-450aa) purified from E. coli | |
| 50 μL | |
| Cell Cycle and Replication, Signal Transduction | |
| 1445 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction