Learn More
csl/RBPJK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-33427
Product Code. 18711704
  • € 629.00 / 0.10mL
Description
csl/RBPJK Polyclonal specifically detects csl/RBPJK in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation (ChIP).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Recombinant protein encompassing a sequence within the center region of human RBPJK. The exact sequence is proprietary.