missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: IPANGLRYGD DYVVGQNVSY MCQPGYTMEL NGSRIRTCTI NGTWSGVMPT CRAVTCPTPP QISNGRLEGT NFDWGFSISY ICSPGYELSF PAVLTCVGN Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 49%.
CUB and Sushi multiple domains 3 (CSMD3) is a gene that encodes a protein with an unknown function. Missense mutations, silent mutations, nonsense mutations, and frameshift deletions are observed in cancers such as lung cancer, skin cancer, and stomach cancer.
Specifications
Specifications
| Antigen | CSMD3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CSMD3 |
| Gene Accession No. | Q7Z407 |
| Gene Alias | 4930500N14Rik; CSMD3; CUB and sushi domain-containing protein 3; CUB and Sushi multiple domains 3; CUB and sushi multiple domains protein 3; Kiaa1894; mKIAA1894 |
| Gene Symbols | CSMD3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
