missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSN3 (CASK) Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 184.00 - € 485.00
Specifications
| Antigen | CSN3 (CASK) |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17720414
![]() |
Invitrogen™
PA5118999 |
200 μL |
€ 485.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17793454
|
Invitrogen™
PA5122061 |
20 μL |
€ 184.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CSN3 (CASK) Polyclonal Antibody for Western Blot, IHC (P)
Kappa-casein (CSN3) stabilizes micelle formation, preventing casein precipitation in milk.Specifications
| CSN3 (CASK) | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Bovine | |
| CSN3 | |
| casein kappa; CASK; CNS10; CSN10; CSN3; CSNK; kappa-casein; KCA | |
| CSN3 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02668, P07498 | |
| 1448, 281728 | |
| Recombinant protein Casein Kappa. The antigen corresponds to amino acid range 24-181 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title