missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSN7b Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-461H
This item is not returnable.
View return policy
Description
CSN7b Polyclonal antibody specifically detects CSN7b in Human samples. It is validated for Western Blot
Specifications
| CSN7b | |
| Polyclonal | |
| Western Blot | |
| Arabidopsis, homolog) subunit 7B, COP9 constitutive photomorphogenic homolog subunit 7B (Arabidopsis), COP9 signalosome complex subunit 7b, CSN7BMGC111077, FLJ12612, Signalosome subunit 7b | |
| The immunogen this antibody was made to, maps to a region between residues 225-264 (C-terminus) of human constitutive photomorphogenic homolog subunit 7B using the TrEMBL entry Q9H9Q2 (GeneID 64708). | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers, Neurotransmission, Signal Transduction, Transcription Factors and Regulators | |
| 64708 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction