missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSNK1G2 Monoclonal Antibody (OTI2F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802864S
This item is not returnable.
View return policy
Description
CSNK1G2 Monoclonal antibody specifically detects CSNK1G2 in Human samples. It is validated for Western BlotSpecifications
| CSNK1G2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P78368 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1455 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI2F2 | |
| Unconjugated | |
| Csnk1g2 | |
| 2810429I12Rik; AI463719; casein kinase 1 gamma 2; casein kinase 1 gamma 2 isoform; casein kinase 1 isoform gamma-2; casein kinase 1, gamma 2; casein kinase I isoform gamma-2; CK Igamma2; CK1G2; CKI gamma 2; CKI gamma2; CKI-gamma 2; CKIgamma2; CK-Igamma2; CKI-gamma2; CKI-gamma-2; CKIgamma-2; Csnk1g2 | |
| Human recombinant protein fragment corresponding to amino acids 163-415 of human CSNK1G2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction