Learn More
Invitrogen™ CSPP1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5103731
Product Code. 16620185
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total CSPP1.
CSPP1 interacts with centrosomes and microtubules and plays a role in cell-cycle progression and spindle organization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CSPP1
Polyclonal
Unconjugated
CSPP1
2310020J12Rik; 4930413O22Rik; AA387550; BC055915; centrosome and spindle pole associated protein 1; Centrosome and spindle pole-associated protein 1; centrosome spindle pole associated protein 1; centrosome/spindle pole-associated protein 2; centrosome/spindle pole-associated protein 3; CSPP; CSPP1; Cspp2; Cspp3; JBTS21; RGD1307055
Rabbit
Affinity chromatography
RUO
211660, 79848
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
B2RX88, Q1MSJ5
CSPP1
A synthesized peptide derived from human CSPP1(Accession Q1MSJ5), corresponding to amino acid residues A1065-D1115.
100 μL
Primary
Human, Mouse