Learn More
Invitrogen™ CSPP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103731
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CSPP1.
CSPP1 interacts with centrosomes and microtubules and plays a role in cell-cycle progression and spindle organization.
Specifications
| CSPP1 | |
| Polyclonal | |
| Unconjugated | |
| CSPP1 | |
| 2310020J12Rik; 4930413O22Rik; AA387550; BC055915; centrosome and spindle pole associated protein 1; Centrosome and spindle pole-associated protein 1; centrosome spindle pole associated protein 1; centrosome/spindle pole-associated protein 2; centrosome/spindle pole-associated protein 3; CSPP; CSPP1; Cspp2; Cspp3; JBTS21; RGD1307055 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 211660, 79848 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B2RX88, Q1MSJ5 | |
| CSPP1 | |
| A synthesized peptide derived from human CSPP1(Accession Q1MSJ5), corresponding to amino acid residues A1065-D1115. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |