missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSRP2BP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111634
This item is not returnable.
View return policy
Description
Immunogen sequence: IYGAKEGGIS RLPAGQATYR TTCQDFRILD RYQTSLPSRK GFRHQTTKFL YRLVGSEDMA VDQSIVSPYT SRILKPYIRR DYETKPPKLQ LLSQIRSHLH Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 94%.
CSRP2 is a protein containing two LIM domains, which are double zinc finger motifs found in proteins of diverse function. CSRP2 and some related proteins are thought to act as protein adapters, bridging two or more proteins to form a larger protein complex. The protein encoded by this gene binds to one of the LIM domains of CSRP2 and contains an acetyltransferase domain. Although the encoded protein has been detected in the cytoplasm, it is predominantly a nuclear protein. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| CSRP2BP | |
| Polyclonal | |
| Unconjugated | |
| KAT14 | |
| 2510008M08Rik; ADA2A-containing complex subunit 2; ATAC component 2 homolog; ATAC2; AU023459; CRP2 binding partner; CRP2 binding protein; CRP2-binding partner; CRP2BP; CSRP2 binding protein; CSRP2-binding protein; Csrp2bp; cysteine and glycine-rich protein 2 binding protein; cysteine-rich protein 2 binding protein; cysteine-rich protein 2-binding protein; D2Ertd473e; D2Wsu131e; dJ717M23.1; E430020F17; Kat14; Lysine acetyltransferase 14; PRO1194 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57325 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H8E8 | |
| KAT14 | |
| Recombinant Protein corresonding to Human CSRP2BP. Recombinant protein control fragment (Product #RP-105448). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction