missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSTB Monoclonal Antibody (OTI2A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813048S
This item is not returnable.
View return policy
Description
CSTB Monoclonal antibody specifically detects CSTB in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CSTB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P04080 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1476 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A7 | |
| Unconjugated | |
| CSTB | |
| AA960480; CPI-B; Cst6; CSTB; Cyb; cystatin B; cystatin B (stefin B); cystatin B protein; cystatin beta; cystatin-B; Cystatin-beta; Epm1; EPM1A; Liver thiol proteinase inhibitor; LOC101123010; PME; stefin B; Stefin-B; stefin-C; Stfb; ULD; Unknown (protein for MGC:134609) | |
| Human recombinant protein fragment corresponding to amino acids 1-98 of human CSTB produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction