missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSTF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566737
This item is not returnable.
View return policy
Description
Immunogen sequence: AKLWEISTGRT LVRYTGAGLS GRQVHRTQAV FNHTEDYVLL PDERTISLCC WDSRTAERRN LLSLGHNNIV RCIVHSPTNP GFMTCSDDFR ARFWYRRS Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
Specifications
| CSTF1 | |
| Polyclonal | |
| Unconjugated | |
| CSTF1 | |
| 1700057K18Rik; AI788832; CF-1 50 kDa subunit; cleavage stimulation factor 50 kDa subunit; Cleavage stimulation factor subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kD; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa; CSTF 50 kDa subunit; Cstf1; CstF50; CstF-50; CstFp50 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1477 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q05048 | |
| CSTF1 | |
| Recombinant Human CSTF1. Recombinant protein control fragment (Product #RP-105348). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction