missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CT110 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99286AF488
This item is not returnable.
View return policy
Description
CT110 Polyclonal antibody specifically detects CT110 in Human samples. It is validated for Western Blot
Specifications
| CT110 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| cancer/testis antigen 110, CT110, FLJ10252, FLJ21048, G patch domain containing 2, G patch domain-containing protein 2, GPATC2, MGC74998, RP11-361K17.1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CT110. | |
| 0.1 mL | |
| Cancer, Immunology | |
| 55105 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction