missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CT110 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 302.00 - € 356.00
Specifications
| Antigen | CT110 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686165
|
Novus Biologicals
NBP2-99286-100ul |
100 μg |
€ 302.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685065
|
Novus Biologicals
NBP2-99286-200ul |
200 μg |
€ 356.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CT110 Polyclonal antibody specifically detects CT110 in Human samples. It is validated for Western BlotSpecifications
| CT110 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Immunology | |
| PBS (pH 7.0) | |
| 55105 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| cancer/testis antigen 110, CT110, FLJ10252, FLJ21048, G patch domain containing 2, G patch domain-containing protein 2, GPATC2, MGC74998, RP11-361K17.1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CT110. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title