missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CT173 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145145
This item is not returnable.
View return policy
Description
Immunogen sequence: VLLGRPQIPQ GSSLGNDIDQ YPVVFRNASD QGSWMQLEML LRKLSDLVWT SDALSDKILE D Antigen sequence identity: Mouse 31%; Rat 31%.
Specifications
| CT173 | |
| Polyclonal | |
| Unconjugated | |
| C20orf173 | |
| C20orf173; chromosome 20 open reading frame 173; dJ477O4.4; Uncharacterized protein C20orf173 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140873 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96LM9 | |
| C20orf173 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 85-145. Recombinant protein control fragment (Product #RP-110238). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction