missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CT45A4 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00441520-B01P-50ug
This item is not returnable.
View return policy
Description
CT45A4 Polyclonal antibody specifically detects CT45A4 in Human samples. It is validated for Western Blot
Specifications
| CT45A4 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| Cancer/Testis Antigen 45-4/6, Cancer/Testis Antigen 45A4/45A6, Cancer/Testis Antigen CT45-4, Cancer/Testis Antigen Family 45 Member A4/A6, Cancer/Testis Antigen Family 45, Member A41, CT45.4, CT45-4, CT45-6 | |
| CT45-4 (AAI53114.1, 1 a.a. - 189 a.a.) full-length human protein. MTDKTEKVAVDPETVFKRPRECDSPSYQKRQRMALLARKQGAGDSLIAGSAMSKAKKLMTGHAIPPSQLDSQIDDFTGFSKDRMMQKPGSNAPVGGNVTSSFSGDDLECRETASSPKSQQEINADIKRKLVKELRCVGQKYEKIFEMLEGVQGPTAVRKRFFESIIKEAARCMRRDFVKHLKKKLKRMI | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AAI53114.1 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 441520 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction