missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTBP1-DT Antibody (OTI2C6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72333PCP
This item is not returnable.
View return policy
Description
CTBP1-DT Monoclonal specifically detects CTBP1-DT in Human samples. It is validated for Flow Cytometry.
Specifications
| CTBP1-DT | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| CTBP1-DT | |
| Recombinant protein fragment corresponding to amino acids 1-186 of human C4orf42 (NP_443093) produced in E.coli. | |
| 0.1 mL | |
| 92070.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2C6 | |
| Flow Cytometry | |
| CTBP1 antisense RNA 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction