missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CtBP1 Mouse anti-Human, Janelia Fluor 646, Clone: 4D6, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-74040JF646
This item is not returnable.
View return policy
Description
CtBP1 Monoclonal antibody specifically detects CtBP1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| CtBP1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| BARS, brefeldin A-ribosylated substrate, CTBP, CtBP1, C-terminal binding protein 1, C-terminal-binding protein 1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.158, MGC104684 | |
| Recombinant human CTBP1 (1-440aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| 1487 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Immunofluorescence | |
| 4D6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction