Learn More
Invitrogen™ CtBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529267
Description
Recommended positive controls: Raji, mouse brain, rat brain. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (83%), Xenopus laevis (96%), Chicken (96%), Rhesus Monkey (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that binds to the C-terminus of adenovirus E1A proteins. This phosphoprotein is a transcriptional repressor and may play a role during cellular proliferation. This protein and the product of a second closely related gene, CTBP2, can dimerize. Both proteins can also interact with a polycomb group protein complex which participates in regulation of gene expression during development. Alternative splicing of transcripts from this gene results in multiple transcript variants.
Specifications
| CtBP1 | |
| Polyclonal | |
| Unconjugated | |
| CTBP1 | |
| 50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; 50-kDaBFA-inducedADP-ribosylatedsubstrate; BARS; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; CTBP; CtBP1; CtBP1-L; CtBP1-S; CtBP3; CtBP3/BARS; C-terminal binding protein 1; C-terminal-binding protein 1; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; D4S115h; D5H4S115; D5H4S115E; MGC104684 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13016, 1487, 29382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.88 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O88712, Q13363, Q9Z2F5 | |
| CTBP1 | |
| Recombinant protein encompassing a sequence within the center region of human CtBP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.