missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTBP2 Antibody (PCRP-CTBP2-1A9), DyLight 650 - C-terminus, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08549C
This item is not returnable.
View return policy
Description
CTBP2 Monoclonal antibody specifically detects CTBP2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| CTBP2 | |
| Monoclonal | |
| DyLight 650 | |
| CtBP2, C-terminal binding protein 2, 10ribeye, C-terminal-binding protein 2 | |
| Recombinant full-length human CTBP2 protein (Uniprot: P56545) | |
| 100 μg | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| 1488 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-CTBP2-1A9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction