missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CtBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521450
This item is not returnable.
View return policy
Description
Recommended positive controls: A375. Predicted reactivity: Mouse (100%), Rat (99%), Xenopus laevis (80%), Xenopus Tropicalis (84%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene produces alternative transcripts encoding two distinct proteins. One protein is a transcriptional repressor, while the other isoform is a major component of specialized synapses known as synaptic ribbons. Both proteins contain a NAD+ binding domain similar to NAD+-dependent 2-hydroxyacid dehydrogenases. A portion of the 3' untranslated region was used to map this gene to chromosome 21q21. 3; however, it was noted that similar loci elsewhere in the genome are likely. Blast analysis shows that this gene is present on chromosome 10.
Specifications
| CtBP2 | |
| Polyclonal | |
| Unconjugated | |
| CTBP2 | |
| AA407280; CTBP2; C-terminal binding protein 2; C-terminal-binding protein 2; D7Ertd45e; gene trap ROSA b-geo 6; Gtrgeo6; Ribeye; RP11-59C5.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1488 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.33 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P56545 | |
| CTBP2 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human CtBP2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction