missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CtBP2 Recombinant Rabbit Monoclonal Antibody (23GB5120), Invitrogen™
GREENER_CHOICE

Product Code. 30089232
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089232 100 μL 100µL
30089235 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089232 Supplier Invitrogen™ Supplier No. MA553850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This gene produces alternative transcripts encoding two distinct proteins. One protein is a transcriptional repressor, while the other isoform is a major component of specialized synapses known as synaptic ribbons. Both proteins contain a NAD+ binding domain similar to NAD+-dependent 2-hydroxyacid dehydrogenases. A portion of the 3' untranslated region was used to map this gene to chromosome 21q21. 3; however, it was noted that similar loci elsewhere in the genome are likely. Blast analysis shows that this gene is present on chromosome 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CtBP2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5120
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CTBP2
Gene Accession No. P56545, P56546, Q9EQH5
Gene Alias AA407280; CTBP2; C-terminal binding protein 2; C-terminal-binding protein 2; D7Ertd45e; gene trap ROSA b-geo 6; Gtrgeo6; Ribeye; RP11-59C5.2
Gene Symbols CTBP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CTBP2 (415-445AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13017, 1488, 81717
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.