missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTBS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554836
This item is not returnable.
View return policy
Description
Immunogen sequence: VSLKDIIDPA FRASWIAQKL NLAKTQYMDG INIDIEQEVN CLSPEYDALT ALVKETTDSF HREIEGSQVT FDVAWSPKNI DRRCYNYTGI ADACDFLFV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
Involved in the degradation of asparagine-linked glycoproteins. Hydrolyze of N-acetyl-beta-D-glucosamine (1-4)N-acetylglucosamine chitobiose core from the reducing end of the bond, it requires prior cleavage by glycosylasparaginase.
Specifications
| CTBS | |
| Polyclonal | |
| Unconjugated | |
| CTBS | |
| 2210401K11Rik; chitobiase; chitobiase, di-N-acetyl-; CTB; CTBdi-N-acetylchitobiase; CTBS; Di-N-acetylchitobiase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1486 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q01459 | |
| CTBS | |
| Recombinant protein corresponding to Human CTBS. Recombinant protein control fragment (Product #RP-92465). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction