missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTBS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112097
This item is not returnable.
View return policy
Description
CTBS Polyclonal Antibody for Western Blot
Involved in the degradation of asparagine-linked glycoproteins. Hydrolyze of N-acetyl-beta-D-glucosamine (1-4)N-acetylglucosamine chitobiose core from the reducing end of the bond, it requires prior cleavage by glycosylasparaginase.
Specifications
| CTBS | |
| Polyclonal | |
| Unconjugated | |
| CTBS | |
| 2210401K11Rik; chitobiase; chitobiase, di-N-acetyl-; CTB; CTBdi-N-acetylchitobiase; CTBS; Di-N-acetylchitobiase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1486 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q01459 | |
| CTBS | |
| A synthetic peptide corresponding to the C-terminus of the Human CTBS. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction