missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTCFL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597639
This item is not returnable.
View return policy
Description
CTCFL Polyclonal Antibody for Western Blot, ELISA
CTCFL is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes. CTCFL is normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation.
Specifications
| CTCFL | |
| Polyclonal | |
| Unconjugated | |
| Ctcfl | |
| BORIS; BORIS like protein; BORIS-like protein; Brother of the regulator of imprinted sites; Cancer/testis antigen 27; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein)-like; CCCTC-binding factor like; CT27; CTCF paralog; CTCF T; CTCFL; CTCF-like protein; CTCF-T; dJ579F20.2; HMG 1L1; HMG-1L1; HMGB1L1; likely orthologue of H. sapiens CCCTC-binding factor-like protein (CTCFL); MGC163358; MGC169105; MGC169106; OTTMUSG00000016680; putative high mobility group protein 1-like 1; putative high mobility group protein B1-like 1; transcriptional repressor CTCFL; Zinc finger protein CTCF-T | |
| Rabbit | |
| Protein G | |
| RUO | |
| 140690, 664799 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| A2APF3, Q8NI51 | |
| Ctcfl | |
| Recombinant Mouse Transcriptional repressor CTCFL protein (1-636AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction