missing translation for 'onlineSavingsMsg'
Learn More

CTDSP2 Antibody, Novus Biologicals™

Product Code. 18648084 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18648084 50 μg 50µL
18698392 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18648084 Supplier Novus Biologicals Supplier No. NBP29736250ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CTDSP2 Polyclonal antibody specifically detects CTDSP2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Spécification

Antigen CTDSP2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.0)
Gene Alias carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2, conserved gene amplified in osteosarcoma, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase 2, NIF2, Nuclear LIM interactor-interacting factor 2, OS4NLI-interacting factor 2, Protein OS-4, PSR2, SCP2EC 3.1.3.16, Small CTD phosphatase 2, Small C-terminal domain phosphatase 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CTDSP2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 10106
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.