missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTDSP2 Antibody (OTI7F5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71552H
This item is not returnable.
View return policy
Description
CTDSP2 Monoclonal specifically detects CTDSP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CTDSP2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CTDSP2 | |
| Full length recombinant protein of human CTDSP2 (NP_005721) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI7F5 | |
| Western Blot, Immunohistochemistry | |
| carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2, conserved gene amplified in osteosarcoma, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase 2, NIF2, Nuclear LIM interactor-interacting factor 2, OS4NLI-interacting factor 2, Protein OS-4, PSR2, SCP2EC 3.1.3.16, Small CTD phosphatase 2, Small C-terminal domain phosphatase 2 | |
| Mouse | |
| Affinity Purified | |
| Protein Phosphatase | |
| 10106.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction