Learn More
Invitrogen™ CTDSP2 Monoclonal Antibody (OTI7F5)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA527044
Product Code. 15956634
  • € 588.00 / 100µL
Description
CTDSP2 Monoclonal Antibody for Western Blot, IHC (P)
CTDSP2 is one of several small CTD phosphatases (SCP). It has been proposed that SCP activity is an evolutionarily conserved transcriptional regulator that acts globally to silence neuronal genes. In addition, expression of phosphatase CTDSP2 mRNA isenhanced in several cancer cell lines with amplification of the CTDSP2 and CDK4 genes, and CTDSP2 coamplifies with CDK4 in some primary sarcomas. CTDSP2 may contribute to the development of a subset of sarcomas.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CTDSP2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O14595
CTDSP2
Full length human recombinant protein of CTDSP2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI7F5
Unconjugated
CTDSP2
AI586070; Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2; conserved gene amplified in osteosarcoma; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2; CTD small phosphatase 2; CTDSP2; D10Ertd73e; NIF2; NLI-interacting factor 2; nuclear LIM interactor-interacting factor 2; nuclear LIM interactor-interacting factor 2 (NLI-interacting factor 2) (Protein OS-4); nuclear LIM interactor-interacting factor 2-like; OS4; OS-4; protein OS-4; PSR2; SCP2; small CTD phosphatase 2; Small C-terminal domain phosphatase 2
Mouse
Affinity chromatography
RUO
10106
-20°C, Avoid Freeze/Thaw Cycles