missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTDSP2 Monoclonal Antibody (OTI7F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808714
This item is not returnable.
View return policy
Description
CTDSP2 is one of several small CTD phosphatases (SCP). It has been proposed that SCP activity is an evolutionarily conserved transcriptional regulator that acts globally to silence neuronal genes. In addition, expression of phosphatase CTDSP2 mRNA isenhanced in several cancer cell lines with amplification of the CTDSP2 and CDK4 genes, and CTDSP2 coamplifies with CDK4 in some primary sarcomas. CTDSP2 may contribute to the development of a subset of sarcomas.Specifications
| CTDSP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14595 | |
| CTDSP2 | |
| Full length human recombit protein of human CTDSP2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7F5 | |
| Unconjugated | |
| CTDSP2 | |
| OS4, PSR2, SCP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10106 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction