missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTDSP2 Monoclonal Antibody (OTI7F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808714S
This item is not returnable.
View return policy
Description
CTDSP2 Monoclonal antibody specifically detects CTDSP2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CTDSP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P57796 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57010 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7F5 | |
| Unconjugated | |
| Cabp4 | |
| AI586070; Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2; conserved gene amplified in osteosarcoma; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2; CTD small phosphatase 2; CTDSP2; D10Ertd73e; NIF2; NLI-interacting factor 2; nuclear LIM interactor-interacting factor 2; nuclear LIM interactor-interacting factor 2 (NLI-interacting factor 2) (Protein OS-4); nuclear LIM interactor-interacting factor 2-like; OS4; OS-4; protein OS-4; PSR2; SCP2; small CTD phosphatase 2; Small C-terminal domain phosphatase 2 | |
| Full length human recombinant protein of human CABP4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction