Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (249936), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1898H
Product Code. 18634743
  • € 706.00 / 0.10mL
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal antibody specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD229/SLAMF3/Lymphocyte Antigen 9
Monoclonal
HRP
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, CyTOF
249936
Western Blot, Flow Cytometry, CyTOF-ready
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9
Mouse myeloma cell line NS0-derived recombinant human CD229/SLAMF3, Lys48-Lys454, Accession # Q9HBG7
0.1 mL
Immunology
4063
Store at 4°C in the dark.
IgG2a