missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTF1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00001489-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MSRREGSLEDPQTDSSVSLLPHLEAKIRQTHSLAHLLTKYAEQLLQEYVQLQGDPFGLPSFSPPRLPVAGLSAPAPSHAGLPVHERLRLDAAALAALPPLLDAVCRRQAELNPRAPRLLRRLEDAARQARALGAAVEALLAALGAANRGPRAEPPAATASAASATGVFPAKVLGLRVCGLYREWLSRTEGDLGQLLPGGSASpecifications
| CTF1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_001330 | |
| CTF1 | |
| CTF1 (NP_001321.1, 1 a.a. ∼ 201 a.a) full-length human protein. | |
| RUO | |
| 1489 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| cardiotrophin 1 | |
| CT-1/CT1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction