missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTGF Monoclonal Antibody (OTI5C10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806800S
This item is not returnable.
View return policy
Description
CTGF Monoclonal antibody specifically detects CTGF in Human samples. It is validated for Western BlotSpecifications
| CTGF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P29279 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1490 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI5C10 | |
| Unconjugated | |
| CCN2 | |
| 2-Nov; betaIG-M2; CCN family member 2; Ccn2; Cellular communication network factor 2; connective tissue growth factor; connective tissue growth factor precursor; Connective tissue growth factor precursor (CTGF) (FISP-12 protein) (Hypertrophic chondrocyte-specific protein 24); connective tissue growth-related protein; CTGF; CTGF protein; CTGRP; fibroblast inducible secreated protein; fibroblast inducible secreted protein; Fisp12; fisp-12; Hcs24; H-CTGF; hypertrophic chondrocyte-specific gene product 24; hypertrophic chondrocyte-specific protein 24; IBP-8; IGF-binding protein 8; IGFBP8; IGFBP-8; Insulin-like growth factor-binding protein 8; MGC102839; NOV2; Protein FISP-12; similar to connective tissue growth factor precursor | |
| Human recombinant protein fragment corresponding to amino acids 27-349 of human CTGF produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction