missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTHRC1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98449FR
This item is not returnable.
View return policy
Description
CTHRC1 Polyclonal specifically detects CTHRC1 in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CTHRC1 | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1 | |
| Produced in rabbits immunized with purified, recombinant CTHRC1 (NP_001258229.1; Met1-Lys230). | |
| 0.1 mL | |
| Primary | |
| Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 115908 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction