missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CtIP Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79810DL594
This item is not returnable.
View return policy
Description
CtIP Polyclonal antibody specifically detects CtIP in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| CtIP | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CtBP-interacting protein, CtIP, DNA endonuclease RBBP8, EC 3.1, RBBP-8, retinoblastoma binding protein 8, Retinoblastoma-binding protein 8, Retinoblastoma-interacting protein and myosin-like, RIMSAE2, Sporulation in the absence of SPO11 protein 2 homolog | |
| The immunogen recognized by this antibody maps to a region between residue 850 and the C-terminus (residue 897) of human CtBP Interacting Protein (Retinoblastoma Binding Protein 8) using the numbering given in entry NP_002885.1 (GeneID 5932). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 5932 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction