missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTLA-4 Antibody (rCTLA4/7219) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20809JF669
This item is not returnable.
View return policy
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CTLA-4 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| Recombinant fragment of human CTLA-4 (exact sequence is proprietary) | |
| 0.1 mL | |
| Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 1493 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rCTLA4/7219 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction