missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTLA-4 Antibody (RM0199-6G10), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12212UV
This item is not returnable.
View return policy
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| CTLA-4 | |
| Monoclonal | |
| DyLight 350 | |
| CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry | |
| RM0199-6G10 | |
| Western Blot, Flow Cytometry | |
| CTLA4 | |
| Mouse recombinant CTLA-4 | |
| 0.1 mL | |
| Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 1493 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction